Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035505.1 | NP_001030582.1 | 3 | 239 | ||
NM_212552.2 | NP_997717.2 | 4 | 328 | |||
GenBank mRNA | BC017744.2 | - | 2 | 164 | ||
BC028282.1 | - | 3 | 199 | |||
BC042036.1 | - | 4 | 324 | |||
BU600037.1 | - | 333 | ||||
DQ225187.1 | - | 4 | 290 |