Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080475.2 | NP_001073944.1 | 4 | 2076 | ||
XM_005246545.3 | XP_005246602.1 | 4 | 2076 | |||
XM_006712521.3 | XP_006712584.1 | 4 | 2076 | |||
XM_011511160.2 | XP_011509462.1 | 5 | 2411 | |||
XM_011511163.2 | XP_011509465.1 | 4 | 2076 | |||
XM_017004071.1 | XP_016859560.1 | 4 | 2076 | |||
XM_017004072.1 | XP_016859561.1 | 3 | 2261 | |||
XM_017004073.1 | XP_016859562.1 | 3 | 1140 | |||
GenBank mRNA | AK123204.1 | - | 2 | 843 | ||
BC151137.1 | - | 3 | 1715 | 9057037 | ||
BX648983.1 | - | 4 | 2015 |