Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080475.2 | NP_001073944.1 | 7 | 2483 | ||
XM_011511160.2 | XP_011509462.1 | 8 | 2818 | |||
XM_017004072.1 | XP_016859561.1 | 6 | 2668 | |||
XM_017004073.1 | XP_016859562.1 | 6 | 1547 | |||
GenBank mRNA | AK057612.1 | - | 1 | 157 | ||
AK123204.1 | - | 5 | 1250 | |||
BC151137.1 | - | 6 | 2122 | 9057037 |