Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014279.2 | NP_001014301.1 | 1 | 797 | ||
GenBank mRNA | AY032883.1 | - | 1 | 703 | ||
BC067873.1 | - | 1 | 545 | 30344324 | ||
BQ670161.1 | - | 1 | 391 |