Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAIR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002288.5 | NP_002279.2 | 3 | 418 | ||
NM_021270.4 | NP_067154.1 | 3 | 418 | |||
XM_011526961.2 | XP_011525263.1 | 2 | 369 | |||
GenBank mRNA | AA133246.1 | - | 3 | 416 | ||
AF013250.1 | - | 3 | 319 | |||
AI262718.1 | - | 3 | 281 | |||
BC069366.1 | - | 3 | 319 | 7262118 | ||
BX106365.1 | - | 3 | 418 |
Gene Symbol: LAIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KU058441.1 | - | 554 |