Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254738.1 | NP_001241667.1 | 5 | 786 | ||
NM_005168.4 | NP_005159.1 | 6 | 693 | |||
GenBank mRNA | AF498969.1 | - | 5 | 508 | ||
AK309442.1 | - | 4 | 367 | |||
AK313224.1 | - | 5 | 760 | |||
BC012513.1 | - | 6 | 648 | 4453777 | ||
BQ440721.1 | - | 5 | 762 | |||
BT006769.1 | - | 5 | 508 | |||
CR542038.1 | - | 5 | 463 | |||
CR542070.1 | - | 5 | 463 | |||
S82240.1 | - | 5 | 487 | |||
X95282.1 | - | 5 | 508 | |||
X97758.1 | - | 5 | 730 |