Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001665.3 | NP_001656.2 | 2 | 349 | ||
XM_005252916.2 | XP_005252973.1 | 2 | 529 | |||
XM_017017719.1 | XP_016873208.1 | 2 | 376 | |||
GenBank mRNA | AF498974.1 | - | 1 | 191 | ||
AY563952.1 | - | 1 | 191 | |||
BC104177.2 | - | 1 | 274 | 40030689 | ||
BC104178.2 | - | 2 | 276 | 40030690 | ||
BI196968.1 | - | 2 | 325 | |||
BT019872.1 | - | 1 | 191 | |||
BX359696.2 | - | 1 | 263 | |||
CB125569.1 | - | 2 | 349 | |||
CR450335.1 | - | 1 | 191 | |||
CR542105.1 | - | 1 | 191 | |||
L11317.1 | - | 1 | 191 | |||
X61587.1 | - | 2 | 320 |