Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040167.1 | NP_001035257.1 | 6 | 852 | ||
NM_001040168.1 | NP_001035258.1 | 6 | 852 | |||
NM_001166355.1 | NP_001159827.1 | 7 | 748 | |||
NM_002304.2 | NP_002295.1 | 7 | 536 | |||
GenBank mRNA | AF193612.1 | - | 6 | 527 | ||
AK096284.1 | - | 7 | 748 | |||
BC014851.1 | - | 7 | 536 | 4453156 | ||
U94354.1 | - | 6 | 525 |