Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177388.1 | NP_001170859.1 | 4, 5 | 513 | ||
NM_002306.3 | NP_002297.2 | 4, 5 | 513 | |||
NR_003225.2 | 2, 3 | 1469 | ||||
XM_011536759.2 | XP_011535061.2 | 5, 6 | 503 | |||
GenBank mRNA | AB006780.1 | - | 4, 5 | 478 | ||
AB209391.1 | - | 3, 4 | 4556 | |||
AF266280.1 | - | 2, 3 | 796 | |||
AK314929.1 | - | 4, 5 | 512 | |||
BC001120.2 | - | 4, 5 | 449 | 3050135 | ||
BC053667.1 | - | 4, 5 | 464 | 6148401 | ||
BC068068.1 | - | 4, 5 | 459 | 6428428 | ||
BG332134.1 | - | 4, 5 | 471 | |||
BQ645629.1 | - | 576 | ||||
BU681070.1 | - | 2, 3 | 501 | |||
BX161455.1 | - | 2, 3 | 1264 | |||
BX641090.1 | - | 2, 3 | 1470 | |||
CD518618.1 | - | 4, 5 | 513 | |||
CR456897.1 | - | 3, 4 | 425 | |||
CR542097.1 | - | 3, 4 | 425 | |||
M35368.1 | - | 4, 5 | 447 | |||
M36682.1 | - | 4, 5 | 458 | |||
M57710.1 | - | 3, 4 | 443 | |||
M64303.1 | - | 3, 4 | 443 | |||
S59012.1 | - | 3, 4 | 425 |