Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177388.1 | NP_001170859.1 | 5 | 591 | ||
NM_002306.3 | NP_002297.2 | 5 | 591 | |||
NR_003225.2 | 3 | 1547 | ||||
XM_011536759.2 | XP_011535061.2 | 6 | 581 | |||
GenBank mRNA | AB006780.1 | - | 5 | 556 | ||
AB209391.1 | - | 4 | 4634 | |||
AF266280.1 | - | 3 | 874 | |||
AK314929.1 | - | 5 | 590 | |||
BC001120.2 | - | 5 | 527 | 3050135 | ||
BC053667.1 | - | 5 | 542 | 6148401 | ||
BC068068.1 | - | 5 | 537 | 6428428 | ||
BG332134.1 | - | 5 | 549 | |||
BU681070.1 | - | 2 | 423 | |||
BX161455.1 | - | 3 | 1342 | |||
BX641090.1 | - | 3 | 1548 | |||
CD518618.1 | - | 5 | 591 | |||
CR456897.1 | - | 4 | 503 | |||
CR542097.1 | - | 4 | 503 | |||
M35368.1 | - | 5 | 525 | |||
M36682.1 | - | 5 | 536 | |||
M57710.1 | - | 4 | 521 | |||
M64303.1 | - | 4 | 521 | |||
S59012.1 | - | 4 | 503 |