Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005567.3 | NP_005558.1 | 2, 3 | 236 | ||
GenBank mRNA | AK055977.1 | - | 2, 3 | 211 | ||
AK057776.1 | - | 2, 3 | 196 | |||
AK293183.1 | - | 2, 3 | 210 | |||
AK295442.1 | - | 2, 3 | 180 | |||
AK301040.1 | - | 2, 3 | 180 | |||
AK301444.1 | - | 2, 3 | 180 | |||
AK307665.1 | - | 2, 3 | 157 | |||
BC002403.2 | - | 2, 3 | 169 | 2822371 | ||
BC002998.1 | - | 2, 3 | 169 | 2822371 | ||
BC015761.1 | - | 2, 3 | 201 | 4858122 | ||
KU178164.1 | - | 1, 2 | 57 | |||
KU178165.1 | - | 1, 2 | 57 | |||
X79089.1 | - | 2, 3 | 188 |