Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGDIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185077.2 | NP_001172006.1 | 6 | 948 | ||
NM_001185078.2 | NP_001172007.1 | 6 | 586 | |||
NM_001301242.1 | NP_001288171.1 | 7 | 606 | |||
NM_001301243.1 | NP_001288172.1 | 5 | 853 | |||
NM_004309.5 | NP_004300.1 | 6 | 718 | |||
NR_125441.1 | 5 | 641 | ||||
XM_011523574.1 | XP_011521876.1 | 5 | 1078 | |||
GenBank mRNA | AF498926.1 | - | 5 | 582 | ||
AK293147.1 | - | 7 | 584 | |||
AK300816.1 | - | 6 | 559 | |||
AK312347.1 | - | 6 | 654 | |||
BC005851.2 | - | 6 | 674 | 2961556 | ||
BC005875.2 | - | 6 | 674 | 2961556 | ||
BC008701.3 | - | 5 | 609 | 2966760 | ||
BC009759.2 | - | 6 | 639 | 3935956 | ||
BC013846.1 | - | 6 | 483 | |||
BC016031.1 | - | 5 | 590 | 3911525 | ||
BC016185.1 | - | 6 | 634 | 4867857 | ||
BC024258.1 | - | 6 | 634 | 3027721 | ||
BC027730.2 | - | 6 | 637 | 4764249 | ||
BC028333.1 | - | 7 | 559 | |||
BC075827.1 | - | 6 | 634 | 6074977 | ||
BC106044.1 | - | 6 | 677 | 6162284 | ||
BP325393.1 | - | 7 | 537 | |||
BT006884.1 | - | 5 | 582 | |||
CR456777.1 | - | 5 | 582 | |||
D13989.1 | - | 5 | 602 | |||
EU794615.1 | - | 5 | 609 | |||
KU177916.1 | - | 5 | 582 | |||
KU177917.1 | - | 6 | 504 | |||
M97579.1 | - | 5 | 582 | |||
X69550.1 | - | 6 | 635 |