Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001683.3 | NP_001001683.1 | 1, 2 | 158 | ||
NM_001305000.1 | NP_001291929.1 | 1, 2 | 158 | |||
GenBank mRNA | AF161414.1 | - | 1, 2 | 133 | ||
BC070377.1 | - | 1, 2 | 138 | 6450545 | ||
BG503911.1 | - | 1, 2 | 139 | |||
HG518163.1 | - | 1, 2 | 153 | |||
HY036468.1 | - | 1, 2 | 158 |