Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003760.4 | NP_001003760.2 | 2 | 1295 | ||
XM_005249107.4 | XP_005249164.1 | 2 | 1490 | |||
XM_017010858.1 | XP_016866347.1 | 2 | 2405 | |||
GenBank mRNA | BC137267.1 | - | 1 | 1180 | 9021264 | |
BC137269.1 | - | 1 | 1180 | 9021266 | ||
EF633513.1 | - | 2 | 1295 |