Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012267.2 | NP_001012267.1 | 3 | 873 | ||
NM_001286969.1 | NP_001273898.1 | 2 | 691 | |||
XM_011518685.2 | XP_011516987.1 | 3 | 884 | |||
XM_011518689.1 | XP_011516991.1 | 3 | 884 | |||
XM_017014715.1 | XP_016870204.1 | 3 | 884 | |||
GenBank mRNA | AK092603.1 | - | 3 | 688 | ||
AK307435.1 | - | 2 | 211 | |||
BC071726.1 | - | 3 | 399 | 5554572 | ||
BX537851.1 | - | 2 | 220 |