Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC403323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122076.1 | 2 | 575 | |||
NR_122077.1 | 2 | 685 | ||||
GenBank mRNA | AK000451.1 | - | 2 | 685 | ||
AK309896.1 | - | 2 | 195 | |||
DB521113.1 | - | 2 | 226 |