Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004311.3 | NP_004302.1 | 5 | 587 | ||
XM_017016260.1 | XP_016871749.1 | 5 | 773 | |||
GenBank mRNA | AF493889.1 | - | 4 | 429 | ||
AK312525.1 | - | 5 | 539 | |||
BC009841.2 | - | 5 | 484 | 3926032 | ||
CR407637.1 | - | 4 | 429 | |||
U07151.1 | - | 4 | 444 |