Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085401.2 | NP_001078870.1 | 3 | 993 | ||
NR_104463.1 | 3 | 993 | ||||
NR_104464.1 | 2 | 615 | ||||
XM_005249112.3 | XP_005249169.1 | 2 | 470 | |||
XM_011514613.2 | XP_011512915.1 | 3 | 607 | |||
XM_017010861.1 | XP_016866350.1 | 2 | 624 | |||
GenBank mRNA | AF429972.1 | - | 2 | 443 | ||
AK124114.1 | - | 2 | 428 | |||
AK301893.1 | - | 2 | 428 | |||
BC047663.1 | - | 3 | 990 | 5266746 | ||
BC066986.1 | - | 2 | 604 | 4827732 |