Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130478.1 | NP_001123950.1 | 5 | 423 | ||
NM_001135655.1 | NP_001129127.1 | 4 | 403 | |||
NM_002347.4 | NP_002338.3 | 4 | 367 | |||
XM_017013413.1 | XP_016868902.1 | 3 | 381 | |||
XM_017013414.1 | XP_016868903.1 | 4 | 428 | |||
GenBank mRNA | AB012293.1 | - | 4 | 347 | ||
AK314139.1 | - | 5 | 423 | |||
BC028894.1 | - | 4 | 393 | 4939118 | ||
BC030192.1 | - | 4 | 327 | 5199071 | ||
BI914261.1 | - | 3 | 208 | |||
BP347868.1 | - | 4 | 331 | |||
CR457034.1 | - | 3 | 201 | |||
DB471677.1 | - | 4 | 367 | |||
DB502453.1 | - | 4 | 416 |