Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033667.2 | NP_001028839.1 | 2 | 471 | ||
NM_001261456.1 | NP_001248385.1 | 2 | 471 | |||
NM_001261457.1 | NP_001248386.1 | 2 | 471 | |||
NM_002348.3 | NP_002339.2 | 2 | 471 | |||
XM_011509548.1 | XP_011507850.1 | 3 | 531 | |||
XM_011509549.1 | XP_011507851.1 | 3 | 531 | |||
XM_011509550.1 | XP_011507852.1 | 3 | 531 | |||
XM_011509552.1 | XP_011507854.1 | 3 | 531 | |||
XM_011509556.1 | XP_011507858.1 | 3 | 531 | |||
XM_011509560.1 | XP_011507862.1 | 3 | 531 | |||
XM_017001297.1 | XP_016856786.1 | 3 | 531 | |||
XM_017001298.1 | XP_016856787.1 | 3 | 531 | |||
XM_017001299.1 | XP_016856788.1 | 3 | 531 | |||
XM_017001300.1 | XP_016856789.1 | 2 | 500 | |||
XM_017001301.1 | XP_016856790.1 | 2 | 371 | |||
XM_017001302.1 | XP_016856791.1 | 3 | 531 | |||
XM_017001303.1 | XP_016856792.1 | 3 | 531 | |||
XM_017001304.1 | XP_016856793.1 | 2 | 371 | |||
GenBank mRNA | AF244129.1 | - | 2 | 387 | ||
AK292048.1 | - | 2 | 371 | |||
AK303403.1 | - | 2 | 371 | |||
AY007142.1 | - | 1 | 193 | |||
BC027920.1 | - | 2 | 307 | 5225850 | ||
BC042905.1 | - | 2 | 358 | |||
BC064485.1 | - | 2 | 358 | |||
BC121049.1 | - | 2 | 406 | 40117946 | ||
BG546913.1 | - | 3 | 605 | |||
DC406206.1 | - | 2 | 471 | |||
L42621.1 | - | 2 | 264 |