Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002349.3 | NP_002340.2 | 4 | 754 | ||
GenBank mRNA | AF011333.1 | - | 4 | 733 | ||
AF064827.1 | - | 4 | 680 | |||
AK310678.1 | - | 4 | 740 |
Gene Symbol: LY75-CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198759.1 | NP_001185688.1 | 4 | 754 | ||
NM_001198760.1 | NP_001185689.1 | 4 | 754 | |||
GenBank mRNA | AY184222.1 | - | 4 | 680 | ||
AY314006.1 | - | 4 | 680 |