Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001343.3 | NP_001001343.2 | 2 | 269 | ||
GenBank mRNA | AK289701.1 | - | 2 | 213 | ||
BC022570.1 | - | 2 | 221 | 4793169 | ||
DA173480.1 | - | 2 | 269 |