Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002356.6 | NP_002347.5 | 1 | 498 | ||
GenBank mRNA | AW163148.1 | - | 1 | 485 | ||
BC013004.2 | - | 1 | 471 | |||
BC028284.1 | - | 1 | 471 | |||
BC068059.1 | - | 1 | 464 | |||
BC089040.1 | - | 1 | 345 | 6378349 | ||
CN345330.1 | - | 1 | 473 | |||
D10522.1 | - | 1 | 456 | |||
M68956.1 | - | 1 | 395 |