Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005908.3 | NP_005899.3 | 17 | 2617 | ||
XM_011531966.2 | XP_011530268.1 | 12 | 1752 | |||
XM_017008203.1 | XP_016863692.1 | 18 | 2603 | |||
XM_017008204.1 | XP_016863693.1 | 16 | 2448 | |||
XM_017008205.1 | XP_016863694.1 | 8 | 3413 | |||
GenBank mRNA | AB209846.1 | - | 17 | 6752 | ||
AK291598.1 | - | 17 | 2605 | |||
AK300015.1 | - | 16 | 2387 | |||
AK312827.1 | - | 17 | 2570 | |||
BC015743.1 | - | 17 | 2607 | 4905535 | ||
EU009130.1 | - | 17 | 2517 | |||
U60337.1 | - | 17 | 2604 |