Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000898.4 | NP_000889.3 | 8 | 1054 | ||
XM_005272608.3 | XP_005272665.1 | 10 | 1154 | |||
XM_017029523.1 | XP_016885012.1 | 10 | 1204 | |||
XM_017029524.1 | XP_016885013.1 | 9 | 1014 | |||
GenBank mRNA | AK294306.1 | - | 9 | 972 | ||
AK298942.1 | - | 8 | 991 | |||
AK312679.1 | - | 8 | 1029 | |||
BC022494.1 | - | 8 | 1000 | 4800208 | ||
M69177.1 | - | 8 | 955 | |||
S62734.1 | - | 8 | 878 |