Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000351.5 | NP_000342.2 | 7 | 1272 | ||
NM_001320750.1 | NP_001307679.1 | 8 | 1496 | |||
NM_001320751.1 | NP_001307680.1 | 9 | 1490 | |||
NM_001320752.1 | NP_001307681.1 | 8 | 1336 | |||
NM_001320753.1 | NP_001307682.1 | 7 | 1114 | |||
NM_001320754.1 | NP_001307683.1 | 7 | 1262 | |||
GenBank mRNA | AK314034.1 | - | 7 | 1212 | ||
BC075030.2 | - | 7 | 1048 | 30915417 | ||
J04964.1 | - | 7 | 1230 | |||
M16505.1 | - | 7 | 1247 |