Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134364.1 | NP_001127836.1 | 2, 3 | 749 | ||
NM_002375.4 | NP_002366.2 | 2, 3 | 749 | |||
NM_030885.3 | NP_112147.2 | 2, 3 | 749 | |||
XM_006713152.3 | XP_006713215.1 | 2, 3 | 742 | |||
XM_017006393.1 | XP_016861882.1 | 2, 3 | 325 | |||
XM_017006394.1 | XP_016861883.1 | 2, 3 | 742 | |||
XM_017006398.1 | XP_016861887.1 | 1, 2 | 242 | |||
XM_017006402.1 | XP_016861891.1 | 2, 3 | 742 | |||
XM_017006405.1 | XP_016861894.1 | 2, 3 | 742 | |||
XM_017006409.1 | XP_016861898.1 | 2, 3 | 742 | |||
XM_017006411.1 | XP_016861900.1 | 2, 3 | 742 | |||
XM_017006412.1 | XP_016861901.1 | 2, 3 | 742 | |||
XM_017006413.1 | XP_016861902.1 | 2, 3 | 742 | |||
XM_017006414.1 | XP_016861903.1 | 2, 3 | 742 | |||
XM_017006415.1 | XP_016861904.1 | 2, 3 | 742 | |||
XM_017006416.1 | XP_016861905.1 | 1, 2 | 242 | |||
GenBank mRNA | AA447222.1 | - | 2, 3 | 380 | ||
AB209377.1 | - | 2, 3 | 309 | |||
AK054577.1 | - | 2, 3 | 744 | |||
BC008715.2 | - | 2, 3 | 313 | 2961360 | ||
BC012794.2 | - | 2, 3 | 313 | 4098499 | ||
BC015149.2 | - | 2, 3 | 313 | 2961360 | ||
BC019229.1 | - | 2, 3 | 279 | |||
BC051843.1 | - | 2, 3 | 294 | |||
BI549723.1 | - | 2, 3 | 701 | |||
M64571.1 | - | 2, 3 | 313 | |||
U19727.1 | - | 2, 3 | 298 |