Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001712.2 | NP_001001712.2 | 3 | 376 | ||
GenBank mRNA | BC040963.1 | - | 3 | 1621 | ||
BC133045.1 | - | 3 | 376 | 40147067 |