Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001712.2 | NP_001001712.2 | 3, 4 | 414 | ||
GenBank mRNA | BC040963.1 | - | 3, 4 | 1659 | ||
BC133045.1 | - | 3, 4 | 414 | 40147067 |