Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000429.2 | NP_000420.1 | 7 | 1129 | ||
XM_005269842.4 | XP_005269899.1 | 8 | 1359 | |||
XM_005269843.4 | XP_005269900.1 | 7 | 1248 | |||
GenBank mRNA | X69078.1 | - | 7 | 945 |