Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001794.3 | NP_001001794.3 | 7 | 681 | ||
XM_005261914.3 | XP_005261971.1 | 8 | 856 | |||
XM_011530692.2 | XP_011528994.1 | 8 | 856 | |||
XM_011530693.2 | XP_011528995.1 | 7 | 635 | |||
XM_017028801.1 | XP_016884290.1 | 6 | 692 | |||
GenBank mRNA | AK054743.1 | - | 6 | 714 | ||
BC031069.1 | - | 7 | 681 | 5296726 | ||
BI758775.1 | - | 8 | 624 | |||
BX363595.2 | - | 7 | 671 |