Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ART3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130016.2 | NP_001123488.1 | 12 | 1249 | ||
NM_001130017.2 | NP_001123489.1 | 10 | 1199 | |||
NM_001179.5 | NP_001170.2 | 11 | 1216 | |||
XM_005262997.1 | XP_005263054.1 | 12 | 1157 | |||
XM_005262999.1 | XP_005263056.1 | 10 | 1091 | |||
XM_005263003.1 | XP_005263060.1 | 8 | 1028 | |||
XM_005263004.1 | XP_005263061.1 | 8 | 1025 | |||
XM_006714220.1 | XP_006714283.1 | 9 | 1061 | |||
XM_011531971.1 | XP_011530273.1 | 9 | 1061 | |||
XM_017008209.1 | XP_016863698.1 | 10 | 1094 | |||
XM_017008210.1 | XP_016863699.1 | 12 | 1602 | |||
GenBank mRNA | AK225860.1 | - | 10 | 1179 | ||
AY550336.1 | - | 8 | 346 | |||
AY550337.1 | - | 7 | 313 | |||
AY550338.1 | - | 5 | 250 | |||
BC008397.1 | - | 12 | 1236 | 4244549 | ||
BC017913.1 | - | 11 | 1203 | 4292261 | ||
BT007249.1 | - | 11 | 1115 | |||
CR541664.1 | - | 11 | 1115 | |||
U47054.1 | - | 10 | 1079 |