Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005921.1 | NP_005912.1 | 14 | 2918 | ||
XM_017009484.1 | XP_016864973.1 | 14 | 5629 | |||
XM_017009485.1 | XP_016864974.1 | 14 | 2357 | |||
GenBank mRNA | AF042838.1 | - | 14 | 2360 |