Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291958.1 | NP_001278887.1 | 20, 21 | 4159 | ||
NM_001301072.1 | NP_001288001.1 | 19, 20 | 4049 | |||
NM_005922.3 | NP_005913.2 | 19, 20 | 4061 | |||
NM_006724.3 | NP_006715.2 | 18, 19 | 3911 | |||
NR_120425.1 | 19, 20 | 4061 | ||||
XM_005266989.2 | XP_005267046.1 | 18, 19 | 4103 | |||
XM_017010869.1 | XP_016866358.1 | 20, 21 | 4485 | |||
GenBank mRNA | AF002715.1 | - | 19, 20 | 3995 | ||
AF116604.1 | - | 7, 8 | 727 | |||
AK124430.1 | - | 2, 3 | 1041 | |||
AK307866.1 | - | 19, 20 | 3915 | |||
BC136276.1 | - | 19, 20 | 3945 | 9020263 | ||
BC143735.1 | - | 19, 20 | 3942 | 9052249 | ||
BC143743.1 | - | 19, 20 | 3941 | |||
BC143750.1 | - | 20, 21 | 4039 | |||
BC146770.1 | - | 18, 19 | 3911 | 9007216 | ||
BE278366.1 | - | 4, 5 | 423 | |||
D86968.2 | - | 18, 19 | 3911 |