Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040002.1 | NP_001035091.1 | 2 | 310 | ||
NM_004527.3 | NP_004518.1 | 1 | 456 | |||
NM_013999.3 | NP_054705.1 | 1 | 456 | |||
XM_011524818.1 | XP_011523120.1 | 1 | 847 | |||
GenBank mRNA | AK291139.1 | - | 1 | 816 | ||
AW410297.1 | - | 2 | 328 | |||
BC069474.1 | - | 1 | 453 | 7262102 | ||
BC069506.1 | - | 1 | 456 | 7262114 | ||
BE898972.1 | - | 310 | ||||
U10492.1 | - | 1 | 456 | |||
U10493.1 | - | 1 | 456 |