Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005924.4 | NP_005915.2 | 2 | 866 | ||
GenBank mRNA | AK309472.1 | - | 4 | 495 | ||
AK313386.1 | - | 2 | 791 | |||
BC017021.1 | - | 2 | 763 | 3917118 | ||
CR536553.1 | - | 2 | 582 | |||
L36328.1 | - | 2 | 611 | |||
X82629.1 | - | 2 | 847 |