Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135247.1 | NP_001128719.1 | 3 | 212 | ||
NM_001135248.1 | NP_001128720.1 | 3 | 231 | |||
NM_002403.3 | NP_002394.1 | 3 | 234 | |||
NM_017459.2 | NP_059453.1 | 3 | 215 | |||
XM_011541476.2 | XP_011539778.1 | 3 | 841 | |||
GenBank mRNA | AK222751.1 | - | 3 | 193 | ||
AK301655.1 | - | 3 | 207 | |||
AU280150.1 | - | 3 | 235 | |||
BC015039.1 | - | 3 | 234 | 3910064 | ||
CR407678.1 | - | 2 | 101 | |||
DN996554.1 | - | 3 | 184 | |||
U19718.1 | - | 2 | 142 |