Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002408.3 | NP_002399.1 | 1 | 1510 | ||
GenBank mRNA | AK056167.1 | - | 2 | 1269 | ||
AK075051.1 | - | 1 | 1108 | |||
AK075199.1 | - | 1 | 1469 | |||
AK293231.1 | - | 2 | 1199 | |||
BC006390.2 | - | 1 | 1486 | 4127135 | ||
BC018912.2 | - | 1 | 1486 |