Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000899.4 | NP_000890.1 | 2 | 237 | ||
NM_003994.5 | NP_003985.2 | 2 | 237 | |||
GenBank mRNA | AF119835.1 | - | 2 | 184 | ||
AF400436.1 | - | 2 | 225 | |||
AF400437.1 | - | 2 | 184 | |||
AK290319.1 | - | 2 | 233 | |||
AK293002.1 | - | 2 | 153 | |||
BC069733.1 | - | 2 | 198 | 7262647 | ||
BC069783.1 | - | 2 | 199 | 7262659 | ||
BC069797.1 | - | 2 | 194 | 7262576 | ||
BC074725.2 | - | 2 | 82 | 30915176 | ||
BC126166.1 | - | 2 | 192 | 8991882 | ||
BC143899.1 | - | 2 | 192 | 9052417 | ||
BX474960.1 | - | 2 | 275 | |||
DC320486.1 | - | 2 | 237 | |||
M59964.1 | - | 2 | 225 |