Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MID1 SIRNA Assays ›
Gene Symbol:
MID1
Gene Name:
midline 1
Gene Aliases:
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
Chromosome Location:
Chr.X: 10445310 - 11111264 on Build GRCh38
UniGene ID:
Hs.27695
Species:
Human
siRNA ID
HSS181108
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MID1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000381.3
NP_000372.1
7
1605
NM_001098624.2
NP_001092094.1
7
1535
NM_001193277.1
NP_001180206.1
7
1394
NM_001193278.1
NP_001180207.1
7
1413
NM_001193279.1
NP_001180208.1
6
1260
NM_001193280.1
NP_001180209.1
6
1146
NM_033289.1
NP_150631.1
7
1491
NM_033290.3
NP_150632.1
7
1397
XM_005274536.1
XP_005274593.1
8
1570
XM_005274537.1
XP_005274594.1
8
1547
XM_006724492.3
XP_006724555.1
8
2169
XM_011545525.2
XP_011543827.1
10
1871
XM_011545526.2
XP_011543828.1
10
1906
XM_011545527.2
XP_011543829.1
8
1816
XM_017029534.1
XP_016885023.1
8
1736
XM_017029535.1
XP_016885024.1
10
1757
XM_017029536.1
XP_016885025.1
9
1833
XM_017029537.1
XP_016885026.1
9
1604
XM_017029538.1
XP_016885027.1
7
1303
XM_017029539.1
XP_016885028.1
7
1465
XM_017029540.1
XP_016885029.1
9
1718
XM_017029541.1
XP_016885030.1
10
1871
XM_017029542.1
XP_016885031.1
9
1718
GenBank mRNA
AF035360.1
-
7
1291
AF041206.1
-
7
1485
AF041208.1
-
7
1605
AF041210.1
-
6
1094
AF230976.1
-
7
1390
AF230977.1
-
7
1390
AF269101.1
-
8
1513
AK025187.1
-
6
575
AK226104.1
-
7
1309
AK291215.1
-
7
1487
AK297205.1
-
8
1401
AK299061.1
-
7
1430
AK315095.1
-
7
1436
AY539962.1
-
6
587
AY539983.1
-
6
630
AY539994.1
-
7
1357
AY539997.1
-
5
504
AY540009.1
-
3
274
AY540010.1
-
3
379
AY540011.1
-
2
336
AY540012.1
-
3
274
AY540013.1
-
2
602
AY540022.1
-
6
1204
AY540023.1
-
6
1090
BC053626.1
-
7
1462
6163881
Y13667.1
-
7
1390
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
negative regulation of microtubule depolymerization
pattern specification process
positive regulation of stress-activated MAPK cascade
protein localization to microtubule
interferon-gamma-mediated signaling pathway
protein binding
microtubule binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-2ddqc:80/100.66.79.182:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline