Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000249.3 | NP_000240.1 | 2 | 339 | ||
NM_001167617.1 | NP_001161089.1 | 2 | 139 | |||
NM_001167618.1 | NP_001161090.1 | 2 | 134 | |||
NM_001167619.1 | NP_001161091.1 | 2 | 139 | |||
NM_001258271.1 | NP_001245200.1 | 2 | 339 | |||
NM_001258274.1 | NP_001245203.1 | 2 | 139 | |||
XM_005265161.1 | XP_005265218.1 | 2 | 229 | |||
XM_005265163.1 | XP_005265220.1 | 2 | 139 | |||
XM_005265164.1 | XP_005265221.1 | 2 | 192 | |||
XM_005265166.1 | XP_005265223.1 | 2 | 147 | |||
XM_011533727.1 | XP_011532029.1 | 2 | 147 | |||
XM_017006449.1 | XP_016861938.1 | 2 | 147 | |||
XM_017006450.1 | XP_016861939.1 | 2 | 192 | |||
XM_017006451.1 | XP_016861940.1 | 2 | 192 | |||
GenBank mRNA | AB209848.1 | - | 2 | 116 | ||
AK295359.1 | - | 2 | 113 | |||
AK298324.1 | - | 2 | 89 | |||
AK298583.1 | - | 2 | 112 | |||
AK302807.1 | - | 2 | 68 | |||
AK311326.1 | - | 2 | 349 | |||
AK311365.1 | - | 2 | 90 | |||
AK312609.1 | - | 2 | 155 | |||
AK316074.1 | - | 2 | 112 | |||
AU127758.1 | - | 2 | 213 | |||
AY517558.1 | - | 2 | 139 | |||
BC006850.1 | - | 2 | 191 | 3451538 | ||
BX648844.1 | - | 2 | 142 | |||
CN414955.1 | - | 2 | 365 | |||
DA517219.1 | - | 2 | 139 | |||
DQ648888.1 | - | 2 | 392 | |||
U07343.1 | - | 2 | 162 | |||
U07418.1 | - | 2 | 182 |