Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR3C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000901.4 | NP_000892.2 | 2 | 945 | ||
NM_001166104.1 | NP_001159576.1 | 2 | 945 | |||
XM_011531975.1 | XP_011530277.1 | 2 | 945 | |||
XM_011531976.2 | XP_011530278.1 | 2 | 1242 | |||
XM_011531977.2 | XP_011530279.1 | 2 | 672 | |||
XM_011531978.2 | XP_011530280.1 | 2 | 945 | |||
XM_017008217.1 | XP_016863706.1 | 2 | 1242 | |||
GenBank mRNA | AB209056.1 | - | 1 | 191 | ||
AJ315514.1 | - | 2 | 796 | |||
AK304318.1 | - | 2 | 947 | |||
BC111758.1 | - | 2 | 649 | 40027353 | ||
M16801.1 | - | 2 | 798 |