Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002422.4 | NP_002413.1 | 8 | 1317 | ||
GenBank mRNA | AK223283.1 | - | 8 | 1235 | ||
AK223291.1 | - | 8 | 1236 | |||
AK313310.1 | - | 8 | 1233 | |||
BC069676.1 | - | 8 | 1237 | 7262578 | ||
BC069716.1 | - | 8 | 1237 | 7262590 | ||
BC074815.2 | - | 8 | 1219 | 30915319 | ||
BC074869.2 | - | 8 | 1219 | 30915468 | ||
BC105954.1 | - | 8 | 1230 | 40033171 | ||
BC107490.1 | - | 8 | 1230 | 40033168 | ||
BC107491.1 | - | 8 | 1230 | 40033170 | ||
X05232.1 | - | 8 | 1217 |