Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004995.3 | NP_004986.1 | 4 | 957 | ||
GenBank mRNA | AK291325.1 | - | 4 | 899 | ||
AK312657.1 | - | 4 | 905 | |||
BC064803.1 | - | 4 | 891 | 6046773 | ||
D26512.1 | - | 4 | 783 | |||
HE978355.1 | - | 4 | 906 | |||
U41078.1 | - | 4 | 672 | |||
X83535.1 | - | 4 | 791 | |||
X90925.1 | - | 4 | 784 | |||
Z48481.1 | - | 4 | 785 |