Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015052.2 | NP_001015052.1 | 2 | 337 | ||
NM_001015054.2 | NP_001015054.1 | 2 | 280 | |||
NM_002434.3 | NP_002425.2 | 3 | 530 | |||
GenBank mRNA | AK308654.1 | - | 2 | 268 | ||
AL520052.3 | - | 3 | 441 | |||
AY258284.1 | - | 2 | 246 | |||
BC014991.1 | - | 2 | 276 | 4909112 | ||
BG823448.1 | - | 2 | 291 | |||
BM796301.1 | - | 2 | 337 | |||
L10752.1 | - | 2 | 198 | |||
M74905.1 | - | 2 | 358 | |||
S51033.1 | - | 1 | 131 |