Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPV17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002437.4 | NP_002428.1 | 6 | 448 | ||
XM_005264326.3 | XP_005264383.1 | 6 | 460 | |||
XM_006712021.3 | XP_006712084.1 | 7 | 600 | |||
XM_017004150.1 | XP_016859639.1 | 7 | 3677 | |||
XM_017004151.1 | XP_016859640.1 | 6 | 496 | |||
XM_017004152.1 | XP_016859641.1 | 6 | 519 | |||
GenBank mRNA | AF088036.1 | - | 6 | 419 | ||
AK093305.1 | - | 4 | 1454 | |||
AK123373.1 | - | 1 | 1158 | |||
BC001115.2 | - | 5 | 439 | 2967261 | ||
BC016289.2 | - | 5 | 382 | |||
BG714998.1 | - | 4 | 259 | |||
BI602840.1 | - | 5 | 698 | |||
KU178195.1 | - | 5 | 392 | |||
KU178196.1 | - | 5 | 357 | |||
KU178198.1 | - | 4 | 276 | |||
X76538.1 | - | 6 | 421 |