Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPV17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002437.4 | NP_002428.1 | 7, 8 | 515 | ||
XM_005264326.3 | XP_005264383.1 | 7, 8 | 527 | |||
XM_006712021.3 | XP_006712084.1 | 8, 9 | 667 | |||
XM_017004150.1 | XP_016859639.1 | 8, 9 | 3744 | |||
XM_017004151.1 | XP_016859640.1 | 7, 8 | 563 | |||
XM_017004152.1 | XP_016859641.1 | 7, 8 | 586 | |||
GenBank mRNA | AK093305.1 | - | 5, 6 | 1521 | ||
BC001115.2 | - | 6, 7 | 506 | 2967261 | ||
BC016289.2 | - | 6, 7 | 449 | |||
BG714998.1 | - | 5, 6 | 326 | |||
KU178195.1 | - | 6, 7 | 459 | |||
KU178196.1 | - | 6, 7 | 424 | |||
KU178198.1 | - | 5, 6 | 343 | |||
X76538.1 | - | 7, 8 | 488 |