Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114357.1 | NP_001107829.1 | 1 | 73 | ||
XM_005263019.3 | XP_005263076.1 | 3 | 426 | |||
XM_011531989.2 | XP_011530291.1 | 2 | 352 | |||
XM_017008236.1 | XP_016863725.1 | 3 | 1819 | |||
XM_017008237.1 | XP_016863726.1 | 4 | 594 | |||
XM_017008238.1 | XP_016863727.1 | 2 | 1603 | |||
XM_017008239.1 | XP_016863728.1 | 2 | 1606 | |||
XM_017008240.1 | XP_016863729.1 | 2 | 355 | |||
XM_017008241.1 | XP_016863730.1 | 3 | 1877 | |||
XM_017008242.1 | XP_016863731.1 | 3 | 1885 | |||
GenBank mRNA | BC127739.1 | - | 1 | 74 | ||
BC141967.1 | - | 7 | 899 | |||
BC141968.1 | - | 1 | 73 | |||
BC150574.1 | - | 1 | 73 | 40127870 |