Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002455.4 | NP_002446.3 | 7 | 1241 | ||
NM_198883.3 | NP_942584.2 | 6 | 1148 | |||
GenBank mRNA | AK313863.1 | - | 8 | 1176 | ||
AX775989.1 | - | 7 | 688 | |||
BC001906.1 | - | 6 | 1050 | 3546178 | ||
BQ003402.1 | - | 2 | 414 | |||
BU552401.1 | - | 2 | 218 | |||
CF529296.1 | - | 2 | 405 |