Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002455.4 | NP_002446.3 | 6 | 1087 | ||
NM_198883.3 | NP_942584.2 | 5 | 994 | |||
GenBank mRNA | AK313863.1 | - | 7 | 1022 | ||
AX775989.1 | - | 6 | 534 | |||
BC001906.1 | - | 5 | 896 | 3546178 | ||
BU552401.1 | - | 1 | 64 | |||
CF529296.1 | - | 3 | 558 |